Medical thermometer, Thermoval® standard
Lieferant: HARTMANN PAUL
9250215
HART9250215EA 10.4 EUR
HART9250215
Medical thermometer, Thermoval® standard
Thermometer Digitalthermometer
Digital thermometer for oral, axillary and rectal measurements of the body temperature.
  • Measuring time: approx. 60 - 90 seconds
  • Easy to read digital display
  • Automatic power-off
  • Displays the last measured value
  • With replaceable long-term battery
  • Acoustic signal indicates when thermometer is ready for operation and when the measurement is completed
Lieferumfang: Supplied with protective cover.