Surgical gloves, latex, sterile, Peha-micron® plus powderfree
Lieferant: HARTMANN PAUL
9425742
HART9425742EA 294 EUR
HART9425742
Surgical gloves, latex, sterile, Peha-micron® plus powderfree
Handschuhe
These disposable gloves are made of thin and soft latex providing great tactile sensitivity, especially at the finger tips. Ideal for microsurgery.
  • Powder-free, smooth surface
  • Easy to put on due to inner polymer coating
  • Modified skin grip surface provides optimal grip
  • Suitable for double-gloving
  • Flexible and tear-resistant, anatomical fit
  • With reinforced cuffs
  • Light brown colour prevents light reflections
Colour: Light brown
EN 455
Standards: EN 455