Capillary tubes with filament
Lieferant: HARVARD APPARATUS
30-0020
HARV30-0020EA 283 EUR
HARV30-0020
Capillary tubes with filament
Pipetten Mikrokapillaren
Capillary tubes, made of high quality borosilicate glass with filaments.
  • Easy filling due to filament
  • Available in standard or thin wall configurations
  • Low tip impedance (less than 50 MΩ)
  • Good for patch clamp pipettes and microinjection needles