Fluosorber filter cannister
Lieferant: HARVARD APPARATUS
34-0415
HARV34-0415EA 660 EUR
HARV34-0415
Fluosorber filter cannister
Filter Gasfilter
Activated charcoal canister that is designed to optimise halogenated gas absorption. Fluosorber is factory tested with an air flow of 60 LPM to ensure resistance is not above 2 cm H₂O.
  • Designed to scavenge 200 g of anesthetic agent
  • Stand-alone for passive scavenging or used with the Fluovac for efficient active scavenging
  • Weighs 1200 g before use and 1400 g when full