Syringe pumps, Pump 11 Elite
Lieferant: HARVARD APPARATUS
70-4500 70-4504 70-4505
HARV70-4500EA 7010 EUR
HARV70-4500 HARV70-4504 HARV70-4505
Syringe pumps, Pump 11 Elite
Pumpen Spritzenpumpen
The Pump 11 Elite series sets a new standard for handling the challenges of new applications. It has served the research community as the work horse infusion pump for micro and macro fluidic applications including MS calibration, reactor dosing, animal drug/nutritional studies, cell injections, electrospinning and more.
  • LCD color touch screen, multi-method programming with alpha/numeric keypad
  • Program simple to complex methods without a PC
  • Enhanced flow performance and force for more challenging applications
  • Advanced connectivity with USB, RS232, RS485 and digital I/O