Intubation cannula
Lieferant: HARVARD APPARATUS
73-2855
HARV73-2855EA 0 EUR
HARV73-2855
Intubation cannula
Kanülen
This intubation cannula is used in chronic studies where the animal needs to be intubated.
  • Outside diameter must fit to the animal’s trachea
  • Luer-adapter for use with any Luer connection
  • Available in several sizes for various species