Dual plate rotators
Lieferant: HARVARD APPARATUS
74-2334
HARV74-2334EA 4100 EUR
HARV74-2334
Dual plate rotators
Schüttler Rotations-/Wippschüttler
Dual plate rotator with variable rotation rates, can be used with the Harvard apparatus 96-well equilibrium dialyser. The rotator speeds up the equilibrium dialysis process by keeping the sample in constant motion thereby ensuring higher reproducibility of results.