Test sieves
Lieferant: HAVERBOECKER
206468758
HAVE206468758EA 299.33 EUR
HAVE206468758
Test sieves
Siebe Analysensiebe
Grid sieve with frame made from aluminium, with parallel cylindrical bars.
Standards: DIN EN 933-3
Lieferumfang: Supplied with serial number, inspection certificate according to DIN EN 10 204 and calibration certificate on request.