Maxi submarine electrophoresis unit, HE 99X
Lieferant: Cytiva
HE99X-15-1.5
HE99X-15-1.5EA 872 EUR
HE99X-15-1.5
Maxi submarine electrophoresis unit, HE 99X
Elektrophorese-Systeme Horizontale Gelelektrophoresesysteme
HE 99X Maxi submarine unit offers a variety of gel lengths and comb sizes so that the gel can be run exactly based on the need.
  • Versatile submarine unit is ideal for any molecular biology lab
  • Run as many as 60 samples at once
Cast gels 15 cm wide × 10, 15, or 20 cm long, with one or two combs. Comb choices range from a preparative comb, for a 1 ml fragment purification sample, to a 30-well comb for high-throughput PCR screening.
Every design detail contributes to reliability and durability with seamless molded high impact plastic buffer tray; banana plugs safe from corrosion; a guard to protect the platinum electrodes; electrode carrier for simple electrode replacement; a rugged molded lid with safety guards to protect against electrical hazard; a large diameter bubble level and leveling feet; strain-relieved high voltage power cords; and red and black details on running trays and buffer chamber for orientation.