Mini LED light box
Lieferant: HEATHROW
120661
HEAT120661EA 333 EUR
HEAT120661
Mini LED light box
Beleuchtung Leuchtkästen
This small light box saves bench space and can be stored in a drawer.
  • Slim light panel
  • LED light source stays cool and has longer life (10000 hours)
  • Low power consumption
Viewing area 125×100 mm is perfect for quick screening of 96-well plates, Petri dishes, electrophoresis gels, slides, and transparencies.
Lieferumfang: Includes six 1.5 V AAA batteries.