Slide staining tray
Lieferant: HEATHROW
HS15951A
HEATHS15951AEA 176 EUR
HEATHS15951A
Slide staining tray
Färbebehälter
Made of a PP and PE blend, this rugged staining tray is a good alternative to metal version. Slide staining tray set, disposable. Provided with four base trays and one lid.
  • Stain, rinse, and dry slides on a single working tray
  • A clean and fresh working surface area every time
  • Compact size and recessed handles for easy transport from work area to sink
  • Two convenient pour spouts for quick disposal of unwanted liquid waste
  • Dark lid protects slides for light sensitive applications
This disposable slide staining tray set reduces possible cross contamination and messy work areas. Up to eight slides fit comfortably onto the base and the deep well holds up to 38 ml.
W×D×H: 305×127×39,5 mm
Lieferumfang: Supplied with four base trays and one lid.