Storage containers, Tubby®
Lieferant: HEATHROW
HS23453
HEATHS23453EA 51 EUR
HEATHS23453
Storage containers, Tubby®
Boxen Lagerboxen
Constructed of high-impact polypropylene, these sturdy storage tubs with lid and dividers are designed to help organize a laboratory.
  • Removable dividers can create as big or as small a compartment as needed
  • The lid snaps in place to secure the contents and for safe transport
  • Perfect for keeping tubes, tips, pipettes, gloves, and other small items organized and out of the way