Digital pocket timer/stopwatch
Lieferant: HEATHROW
HS24490D
HEATHS24490DEA 41.1 EUR
HEATHS24490D
Digital pocket timer/stopwatch
Timer Timer, digital
Four independent timing channels with visual and audible alarms.
  • Simply move the program switch to use as a stopwatch, a countdown timer (up to 100 hours), or a clock
  • Timing capacity: 100 hours
  • Resolution: ¹/₁₀₀ seconds
  • Count direction: up/down
Lieferumfang: Includes spring clip and magnetic back.