Racks for upright freezers
Lieferant: HEATHROW
HS28622
HEATHS28622EA 390 EUR
HEATHS28622
Racks for upright freezers
Gestelle Kryogestelle
Stainless steel.
For upright freezers and used for 15 and 50 ml tube box storage. Corrosion-resistant construction with (2×3) rack spaces.