Pipette controller, RF3000®
Lieferant: HEATHROW
HS3000
HEATHS3000EA 549 EUR
HEATHS3000
Pipette controller, RF3000®
Pipetten Pipettierhelfer
This microprocessor controlled pipette controller can be used with glass and plastic pipettes from 1 to 100 ml.
  • Long-life li-ion battery, with quick charge-up to 20 hours continuous use
  • Universal grip and soft coated triggers with direction indicators, for right or left handed users
  • Ergonomically balanced in the hand with a loaded pipette
  • Bright LEDs indicate three operation modes: high, low, and gravity
  • Toggle operation by pressing the large mode button
  • Low battery indicator; can be recharged when in use
  • Five-year warranty unit
  • One-year warranty rechargeable battery
An adapter can be fitted to the nose cone to stabilise small volume and thinner pipettes.
Lieferumfang: Supplied with charging stand, wall-mountable/magnetic holder, nozzle adapter for use with small pipettes, Li-Ion rechargeable battery, three replacement 0,45 µm filters, five nose cones (blue, black, yellow, red, grey), and low voltage micro-USB connector (5VDC) and approx. 6 ft (183 cm) cable connected to hi-efficiency double-insulated AC wall outlet adapter 100 to 240 V, 50/60 Hz with four interchangeable plugs.