Cell spreaders, stainless steel
Lieferant: HEATHROW
HS86655
HEATHS86655EA 23.1 EUR
HEATHS86655
Cell spreaders, stainless steel
Schaber Zellschaber
Stainless steel spreader for use with 90 mm plates. Smooth triangular end makes spreading easy - the upturned end will not cut into agar.