Deioniser, mixed-bed, Aquademat
Lieferant: GLASWARENFABRIK KARL HECHT
40075010
HECH40075010EA 393 EUR
HECH40075010
Deioniser, mixed-bed, Aquademat
Wasseraufbereitungssysteme Ionenaustauscher
Ion exchanger for purified water.
  • Blue lightproof cartridge avoids growth of algae
Standards: Conforms with DAB, EuAB.
Lieferumfang: Supplied with conductivity meter with inlet hose, outlet hose and wall holder (hose connections R19 mm).