Haematocrit sealing material
Lieferant: GLASWARENFABRIK KARL HECHT
40566010
HECH40566010EA 2.7 EUR
HECH40566010
Haematocrit sealing material
Versiegeln Versiegelungsmaterial
This sealing material prevents the draining of blood samples. For sealing several hundred micro hematocrit capillaries.