Microscopy slide according to Undritz
Lieferant: GLASWARENFABRIK KARL HECHT
40619010
HECH40619010EA 3.8 EUR
HECH40619010
Microscopy slide according to Undritz
Objektträger Microscopy Control Slides
This microscopy slide is designed for blood smears according to Undritz.
  • Polished surface
  • Clear glass