Instruments/Medicine trays, with Plexiglas® stand
Lieferant: GLASWARENFABRIK KARL HECHT
41105010
HECH41105010EA 63.8 EUR
HECH41105010
Instruments/Medicine trays, with Plexiglas® stand
Schalen/Schüsseln/Töpfe Instrumentenschalen
Made of melamin/Plexiglas®
  • Each row of holes has an area for inscription (pencil notes can easily be removed by rubber or moist cloth)
  • The free space at the side is provided for other medicines, instruments etc.
  • Without cups
Consisting of plastic tray of melamine, white, rack of plexiglas® for 20 medicine cups.