Bunsen burner
Lieferant: GLASWARENFABRIK KARL HECHT
41385010
HECH41385010EA 72.7 EUR
HECH41385010
Bunsen burner
Heizgeräte Gasbrenner
All gas bunsen burner with air regulation, needle valve and adjustable nozzle.
  • Height of approx. 160 mm
  • Tube Ø of approx. 13 mm