Gas burners, Teclu
Lieferant: GLASWARENFABRIK KARL HECHT
41393001
HECH41393001EA 76.2 EUR
HECH41393001
Gas burners, Teclu
Heizgeräte Gasbrenner
Gas burners according to Teclu, with stopcock and pilot flame.
  • Tube ~13 mm Ø
  • Height ~160 mm
  • Base ~80 mm Ø