Digital interval timer
Lieferant: GLASWARENFABRIK KARL HECHT
41977020
HECH41977020EA 17.7 EUR
HECH41977020
Digital interval timer
Timer Timer, digital
Digital interval timer with white casing and magnetic back.
  • LCD display, 4 digits showing minutes and seconds
  • Programmable up to 99 minutes and 59 seconds
  • Counts down in seconds
  • Alarm sounds for 30 seconds when countdown reaches zero
  • Memory recall function
  • Individual setting for minutes and seconds