Analogue stopwatch
Lieferant: GLASWARENFABRIK KARL HECHT
41978010
HECH41978010EA 258 EUR
HECH41978010
Analogue stopwatch
Timer Stoppuhren, analog
Solid metal stopwatch with 7 jewels.
  • Graduation 0 to 60 in 1/5 seconds and 1/100 minutes
  • Timing capacity: Up to 30 minutes
  • Impact, dust and water resistant