Analogue stopwatch, bench top
Lieferant: GLASWARENFABRIK KARL HECHT
41979010
HECH41979010EA 141 EUR
HECH41979010
Analogue stopwatch, bench top
Timer Stoppuhren, analog
Electronic stopclock for bench top use or for wall mounting.
Timing capacity: 0 to 60 seconds, 0 to 60 minutes (scale outside), 0 to 100 / 100 minutes (scale inside).
Lieferumfang: Including battery (AA) 1,5 V.