Laboratory tray set
Lieferant: GLASWARENFABRIK KARL HECHT
42000016
HECH42000016EA 18.7 EUR
HECH42000016
Laboratory tray set
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Complete laboratory tray set consists of plastic tray, Plexiglas® stand and 5 wide neck amber glass bottles (30 ml).