Magnetic Stirrer, Fully Electronic
Lieferant: GLASWARENFABRIK KARL HECHT
42072012
HECH42072012EA 1560 EUR
HECH42072012
Magnetic Stirrer, Fully Electronic
Rührer Magnetrührer
Square CERAN® heating plate fully electronic control of the speed and the heating plate temperature, with digital display and timer function.
  • Possibility of connecting a PT-100 temperature sensor
  • Heating plate dimensions: approx. 135×135 mm
Motor: Gap engine.