Instrument trays with lid
Lieferant: GLASWARENFABRIK KARL HECHT
42915003
HECH42915003EA 28.4 EUR
HECH42915003
Instrument trays with lid
Schalen/Schüsseln/Töpfe Instrumentenschalen
Made of stainless steel.
  • Resistant to hot-air sterilization up to +180 °C
  • Excellent chemical resistance
  • With knob lid