Scheidetrichter, konisch
Lieferant: GLASWARENFABRIK KARL HECHT
42950024
HECH42950024EA 41.4 EUR
HECH42950024
Scheidetrichter, konisch
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, with solid glass or interchangeable PTFE key and plastic stopper.
  • Large labeling field
  • Excllent temperature resistance
  • Excllent chemical resistance
Standards: DIN ISO4800