Scheidetrichter, birnenförmig
Lieferant: GLASWARENFABRIK KARL HECHT
42970066
HECH42970066EA 74.4 EUR
HECH42970066
Scheidetrichter, birnenförmig
Trichter Scheidetrichter
Separating funnels, made of borosilicate glass, pear shaped, stopcock with solid glass plug.
  • Without graduations
  • Excellent chemical resistance
  • Excellent temperature resistance