Max/min thermometer
Lieferant: GLASWARENFABRIK KARL HECHT
43647010
HECH43647010EA 12.5 EUR
HECH43647010
Max/min thermometer
Thermometer Analoge Thermometer
Indoor/outdoor thermometer in beige plastic casing.
  • Case with push button magnet
  • Environmentally friendly capillary filling
  • With roof
Dimensions: 85×45×232 mm