Hydrometer according to Richter and Tralles
Lieferant: GLASWARENFABRIK KARL HECHT
46175010
HECH46175010EA 22.7 EUR
HECH46175010
Hydrometer according to Richter and Tralles
Aräometer
Hydrometer designed without thermometer.
  • Reference temperature: 20 °C (68 °F)
Verpackung: In a protective plastic tube.