Children's Milk Bottle, Wide Neck
Lieferant: GLASWARENFABRIK KARL HECHT
51866010
HECH51866010EA 34.5 EUR
HECH51866010
Children's Milk Bottle, Wide Neck
Flaschen Glasflaschen
Children's milk bottle, wide mouth, with graduations from 20 to 200 ml.
  • Borosilicate glass 3.3
  • Graduated blue
  • Flat floor
  • Strong rolled edge