Rectangular cover glasses
Lieferant: GLASWARENFABRIK KARL HECHT
40990127 40990166 41000133 92099001145 40990157 92099001280 41000142 40990151 40990118 40990130 92099005095
525-1310EA 58.7 EUR
525-1310 HECH40990166 HECH41000133 HECH92099001145 HECH40990157 HECH92099001280 HECH41000142 HECH40990151 HECH40990118 HECH40990130 HECH92099005095
Rectangular cover glasses
Deckgläser
Cover glass D 263® M made of borosilicate glass.
  • Free from colour distortion and highly transparent
  • Excellent quality, low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
Standards: ISO 8255