Square cover glasses
Lieferant: GLASWARENFABRIK KARL HECHT
92101401240
HECH92101401240EA 0 EUR
HECH92101401240
Square cover glasses
Deckgläser
Square cover glasses are made up of colourless borosilicate glass.
  • Low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals
  • Refractive index finely adapted to microscopes
Standards: ISO 8255