Electronic Overhead Stirrers, Hei-TORQUE Silver Package
Lieferant: Heidolph Instruments GmbH & Co.KG
501-61019-00
HEID501-61019-00EA 2287 EUR
HEID501-61019-00
Electronic Overhead Stirrers, Hei-TORQUE Silver Package
Rührer Rührwerke
Hei-TORQUE Silver package with the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory.
  • Hei-TORQUE Expert 100
  • Telescope stand
  • Clamp
Verpackung: Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.