Hei-TORQUE Overhead Stirrers, Gold 2 Package
Lieferant: Heidolph Instruments GmbH & Co.KG
501-62029-00
HEID501-62029-00EA 2844 EUR
HEID501-62029-00
Hei-TORQUE Overhead Stirrers, Gold 2 Package
Rührer Rührwerke
In order to provide the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory, we have added packages to the Hei-TORQUE product group. Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.
Lieferumfang: Hei-TORQUE Ultimate 200 comes with telescope stand and clamp.