Hei-PLATE Sensor Advanced Package Core Magnetic Stirrers
Lieferant: Heidolph Instruments GmbH & Co.KG
506-11157-00
HEID506-11157-00EA 1446 EUR
HEID506-11157-00
Hei-PLATE Sensor Advanced Package Core Magnetic Stirrers
Rührer Magnetrührer
Hei-PLATE Sensor Advanced Package Core.
  • Includes:
  • The Hei-PLATE Mix 'n' Heat Core magnetic stirrer (145 mm diameter) - fast and precise heating - without overshoot - for sample-friendly work
  • EKT Hei-Con electronic contact thermometer incl. holder
  • silicone protective cover for longer lasting protection of your magnetic stirrer