Hei-VAP Industrial - All-round Package Rotary Evaporators
Lieferant: Heidolph Instruments GmbH & Co.KG
518-41100-00
HEID518-41100-00EA 0 EUR
HEID518-41100-00
Hei-VAP Industrial - All-round Package Rotary Evaporators
Verdampfer Rotationsverdampfer
Hei-VAP Industrial - All-round Package.
  • This packages includes:
  • Hei-VAP industrial model with glassware R - Recommended for all standard distillations
  • Base cart
  • Tubing set with 16 meter piece incl. hose clamps
  • Vacuum pump Hei-VAC valve industrial
  • Chiller Hei-CHILL 3000
  • 40 L - Kryo 30 thermal fluid