Chillers, Hei-CHILL 1100 Pro
Lieferant: Heidolph Instruments GmbH & Co.KG
591-01660-00
HEID591-01660-00EA 5244 EUR
HEID591-01660-00
Chillers, Hei-CHILL 1100 Pro
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Hei-CHILL 1100 Pro with a cooling capacity of up to 1100 W allows an effective and powerful cooling of every application, even at low temperatures, or the supply of multiple evaporation systems by a recirculation chiller.
  • Touchscreen controller with glass surface and integrated level indicator
  • Soft running noises for a trouble-free work with speed-controlled fans
  • For applications from –10 to +40 °C with a temperature constancy of +/– 0,5 °C, reliable cooling for different workflows
It has the highest performance for achieving the set target values quickly and for keeping temperatures constant even when using high-polar solvents or during continuous evaporation processes. It is environmentally friendly by using natural refrigerants (R-290) and an efficient cooling system with low-energy fans.
The chiller consists of RS-232 connection option for activating device via a Hei-VAP Expert Control or Ultimate Control or via the Heidolph laboratory software Process solutions.