HelixMark® Peristaltic Pump Tubing on Spool
Lieferant: Freudenberg Medical Europe GmbH
49402092 49300596 49300599 49300600 49451072
HELI49402092EA 1030 EUR
HELI49402092 HELI49300596 HELI49300599 HELI49300600 HELI49451072
HelixMark® Peristaltic Pump Tubing on Spool
Schläuche Transferschlauch
HelixMark® peristaltic pump tubing is designed and manufactured for use with peristaltic pumps and other applications with higher effective forces. A special production process designed to meet these requirements ensures that the products provide greater stability and capacity to withstand stresses. The range offers sizes suitable for use with all commonly used pump types.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Seamless batch
More Product Information