Lids, Silicone Gaskets, and Screw Caps for Standard Cells
Lieferant: HELLMA
01000110VE1072 01000150-VE572 011600-VE10734 011601-VE10734 011650-VE1072 011651-VE1072 010-001-00-10 010-001-1-10
HELL01000110VE1072EA 23 EUR
HELL01000110VE1072 HELL01000150-VE572 HELL011600-VE10734 HELL011601-VE10734 HELL011650-VE1072 HELL011651-VE1072 HELL010-001-00-10 HELL010-001-1-10
Lids, Silicone Gaskets, and Screw Caps for Standard Cells
Küvetten Spektralphotometer-Küvetten
An assortment of lids, silicone gaskets, and screw caps designed for use with standard Hellma® cells.
  • Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers
  • Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism
  • Hellma® precision cells can be easily cleaned and reused and thus remain in use for a long time
Open screw caps are suitable for use with cell types 117.100, 117.100F, 117.104, 117.104F.
Closed screw caps are suitable for use with cell types 117.200, 117.200F, 117.204, 117.204F.
Replacement gaskets are suitable for use with cell types 117.100, 117.100F, 117.104, 117.104F, 117.200, 117.200F, 117.204, 117.204F.