TrayCell® Spectrophotometer Cuvettes
Lieferant: HELLMA
105810-A3-V1-4
HELL105810-A3-V1-4EA 0 EUR
HELL105810-A3-V1-4
TrayCell® Spectrophotometer Cuvettes
Küvetten Spektralphotometer-Küvetten
The TrayCell is a fiber-optic ultra-micro measuring cell designed for the UV/Vis analysis of DNA/RNA and proteins. The
dimensions of the TrayCell are equivalent to a standard cuvette in order to work in most spectrophotometers.
  • Suitable for almost any current spectrophotometers
  • Ideally suited for very small
  • During measurements, the TrayCell shows excellent reproducibility
Trouble-free measurement of the sample at different optical path lengths simply by exchanging the cap (caps with dilution factors: 5, 10, 50 and 100).
Fast and simple cleaning of the optics before measuring the next sample - the TrayCell remains in the cell holder. Samples can be reused after the measurement simply by pipetting them off.
Lieferumfang: TrayCell, two caps with an optical path length of 0,2 and 1,0 mm, two adapters for a center height of 15 and 20 mm, screwdriver for center height adapter, premium storage box.