Cryo Gloves and Protective Wear, Thermo Fisher Scientific
Lieferant: Thermo Scientific
4426
HERA4426EA 0 EUR
HERA4426
Cryo Gloves and Protective Wear, Thermo Fisher Scientific
Handschuhe
Keep your hands protected from ultra-low and cryo temperatures with these highly insulated cryo gloves.
  • Comfortable, lightweight, and breathable protection
  • Available in a variety of sizes
Standards:
cULus and CE.
Verpackung:
One pair of cryo gloves provided.