Aqua-Tec™ Water Preservation Cell
Lieferant: Thermo Scientific
50109579
HERA50109579EA
445
EUR
HERA50109579
Aqua-Tec™ Water Preservation Cell
Reinigungsmittel
Wasserbadschutzmittel
Aqua-Tec water preservation cell, actively fights against unwanted microbial growth.
Can be placed into the water reservoir of any CO₂ incubator or water bath to reduce the risk of contamination.
Bestellinformation: This product is only available in the UK, Ireland, Germany, France, Austria, Switzerland, Sweden, Finland and Iceland.