Temperature chart recorder
Lieferant: Thermo Scientific
6383-6
HERA6383-6EA 2830 EUR
HERA6383-6
Temperature chart recorder
Diagrammschreiber Diagrammschreiber
These recorders are ideal when ultra-low temperature freezers need to meet validation and regulatory requirements.
  • Record seven-day history on 150 mm charts with pressure sensitive chart paper
  • No inking is required