Blood Bank Refrigerators, TSB
Lieferant: Thermo Scientific
TSB140REGBV TSB250REGBV TSB400REGBV TSB700REGBV TSB1500REGBV
HERATSB140REGBVEA 6070 EUR
HERATSB140REGBV HERATSB250REGBV HERATSB400REGBV HERATSB700REGBV HERATSB1500REGBV
Blood Bank Refrigerators, TSB
Kühlschränke
The Thermo Scientific TSB series blood bank refrigerators are designed to meet the strict requirements for storage of blood and blood related products in full compliance with 93/42/CE - class IIa regulation.
  • Touchscreen digital controller with onboard datalogging
  • Whisper quite operation <50 dBA
  • Triple pane glass doors with steel cabinet material
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms
  • Low energy consumption with real time energy monitoring
  • Low heat output
  • Electronic lock with password protection
These refrigerators have casters and rear access port.
Warranty: 36 months
Smart View Compatibility: Smart-Vue Pro compatible
Standards: Certified by 93/42/CEE, 2014/30/UE, 2014/35/UE, 2014/53/UE (only if wireless option), 2011/65/UE.