[EN]TSX REF GLASS 12cf 230v/50Hz EU PLUG 1 * 1 ST
Lieferant: Thermo Scientific
TSX series high-performance lab refrigerator, TSX1205GV, EU plug, Segment: Laboratory/pharmaceutical, Capacity: 326 L, Defrost: Automatic, Shelves/drawers: 4 full size shelves