[EN]TSX REF SLIDING DOOR 120V/60HZ 1 * 1 ST
Lieferant: Thermo Scientific
TSX series high-performance lab refrigerator, TSX4505GA, NEMA 5 - 15 plug, Segment: Laboratory/pharmaceutical, Capacity: 1297 L, Defrost: Automatic, Shelves/drawers: 8 full size shelves