[EN]TSX REFRIGERATOR DOUBLE GLASS50CF230 1 * 1 ST
Lieferant: Thermo Scientific
TSX series high-performance lab refrigerator, TSX5005GV, CEE 7/7 plug, Segment: Laboratory/pharmaceutical, Capacity: 1297 L, Defrost: Automatic, Shelves/drawers: 8 full size shelves